Läs mer
Olivetolic Acid, TRC
Produktkod. 30010379
Change view
Kvantitet:
100 mg
500 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010379 100 mg
30003553 500 mg
30005038 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30010379 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO533005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 491-72-5
Molekylformel C12 H16 O4
Molekylvikt (g/mol) 224.25
Synonym β-Resorcylic acid, 6-pentyl- (7CI,8CI), 2,4-Dihydroxy-6-pentylbenzoic acid, Allazetolcarboxylic acid, Olivetolcarboxylic acid, Olivetolic acid
IUPAC-namn 2,4-dihydroxy-6-pentylbenzoic acid
LEDER CCCCCc1cc(O)cc(O)c1C(=O)O
Specifikationer
Kemiskt namn eller material 2,4-dihydroxy-6-pentyl-benzoic acid
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C12H16O4/c1-2-3-4-5-8-6-9(13)7-10(14)11(8)12(15)16/h6-7,13-14H,2-5H2,1H3,(H,15,16)
Formel vikt 224.1049
Procent renhet >95%