Läs mer
Olmesartan Lactone Impurity, TRC
Produktkod. 30137648
Change view
Kvantitet:
5 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137648 50 mg
17959254 5 mg
17979634 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137648 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO55003050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 849206-43-5
Molekylformel C24 H24 N6 O2
Molekylvikt (g/mol) 428.49
Synonym 4H-Furo[3,4-d]imidazol-4-one, 3,6-dihydro-6,6-dimethyl-2-propyl-3-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-, 4H-Furo[3,4-d]imidazol-4-one, 3,6-dihydro-6,6-dimethyl-2-propyl-3-[[2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]- (9CI), 6,6-Dimethyl-2-propyl-3-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-3,6-dihydro-4H-furo[3,4-d]imidazol-4-one, 1-{[2'-(1H-Tetrazol-5-yl)biphenyl-4-yl]methyl}-4,4-dimethyl-2-propyl-1H-furo[3,4-d]imidazol-6(4H)-one, Olmesartan Medoxomil RC A (USP)
IUPAC-namn 6,6-dimethyl-2-propyl-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]furo[3,4-d]imidazol-4-one
LEDER CCCc1nc2c(C(=O)OC2(C)C)n1Cc3ccc(cc3)c4ccccc4c5nnn[nH]5
Specifikationer
Kemiskt namn eller material 1-{[2'-(1H-Tetrazol-5-yl)biphenyl-4-yl]methyl}-4,4-dimethyl-2-propyl-1H-furo[3,4-d]imidazol-6(4H)-one
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C24H24N6O2/c1-4-7-19-25-21-20(23(31)32-24(21,2)3)30(19)14-15-10-12-16(13-11-15)17-8-5-6-9-18(17)22-26-28-29-27-22/h5-6,8-13H,4,7,14H2,1-3H3,(H,26,27,28,29)
Formel vikt 428.1961
Procent renhet >95%