Läs mer
Olmesartan, LoGiCal
Produktkod. 30463754
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30463754 10 mg
1 options
Produktkod. 30463754 Leverantör LoGiCal Leverantörsnummer LGCFOR1317.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Olmesartan Acid
CAS 144689-24-7
Analyt- eller komponentnamn Olmesartan Acid
Frakt skick Room Temperature
Molekylformel C24 H26 N6 O3
Certifikat och efterlevnad ISO 9001
Synonym Olmesartan Medoxomil Imp. A (EP), 4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-(1H-tetrazol-5-yl)biphenyl-4-yl]methyl]-1H-imidazole-5-carboxylic Acid, Olmesartan, Olmesartan Acid, Olmesartan Medoxomil Impurity A
InChI formel InChI=1S/C24H26N6O3/c1-4-7-19-25-21(24(2,3)33)20(23(31)32)30(19)14-15-10-12-16(13-11-15)17-8-5-6-9-18(17)22-26-28-29-27-22/h5-6,8-13,33H,4,7,14H2,1-3H3,(H,31,32)(H,26,27,28,29)
Lösningstyp Neat
LEDER CCCc1nc(c(C(=O)O)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4)C(C)(C)O
IUPAC-namn 5-(2-hydroxypropan-2-yl)-2-propyl-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylic acid
Industry Type Clinical Testing
Molekylvikt (g/mol) 446.5
Kvantitet 10 mg
Formel vikt 446.2066 g/mol