Läs mer
Olmesartan Methyl Ester, TRC
Produktkod. 30412164
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30412164 10 mg
1 options
Produktkod. 30412164 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO55003510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Olmesartan Methyl Ester
CAS 1347262-29-6
Molekylformel C25H28N6O3
Kvantitet 10 mg
Synonym 4-(1-Hydroxy-1-methylethyl)-2-propyl-1-[[2'-(2H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl]methyl]-1H-imidazole-5-carboxylic Acid Methyl Ester
InChI formel InChI=1S/C25H28N6O3/c1-5-8-20-26-22(25(2,3)33)21(24(32)34-4)31(20)15-16-11-13-17(14-12-16)18-9-6-7-10-19(18)23-27-29-30-28-23/h6-7,9-14,33H,5,8,15H2,1-4H3,(H,27,28,29,30)
LEDER CCCc1nc(c(C(=O)OC)n1Cc2ccc(cc2)c3ccccc3c4nnn[nH]4)C(C)(C)O
IUPAC-namn methyl 5-(2-hydroxypropan-2-yl)-2-propyl-3-[[4-[2-(1H-tetrazol-5-yl)phenyl]phenyl]methyl]imidazole-4-carboxylate
Molekylvikt (g/mol) 460.53
Formel vikt 460.2223