Läs mer
Olopatadine Isopropyl Ester, TRC
Produktkod. 30411097
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30411097 50 mg
30424995 10 mg
30428497 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30411097 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO57502550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1206456-44-1
Molekylformel C24 H29 N O3
Molekylvikt (g/mol) 379.49
Synonym (11Z)-11-[3-(Dimethylamino)propylidene]-6,11-dihydrodibenz[b,e]oxepin-2-acetic acid 1-methylethyl ester
IUPAC-namn propan-2-yl 2-[(11Z)-11-[3-(dimethylamino)propylidene]-6H-benzo[c][1]benzoxepin-2-yl]acetate
LEDER CC(C)OC(=O)Cc1ccc2OCc3ccccc3\C(=C\CCN(C)C)\c2c1
Specifikationer
Kemiskt namn eller material Olopatadine Isopropyl Ester
Kvantitet 50 mg
InChI formel InChI=1S/C24H29NO3/c1-17(2)28-24(26)15-18-11-12-23-22(14-18)21(10-7-13-25(3)4)20-9-6-5-8-19(20)16-27-23/h5-6,8-12,14,17H,7,13,15-16H2,1-4H3/b21-10-
Formel vikt 379.2147