Läs mer
EVOS™ Olympus 10X Objective, super-apochromat, coverslip-corrected
Produktkod. 15532215
Change view
Klicka för att se tillgängliga alternativ
:
Magnification: 10X; Numerical Aperture: 0.4; Working distance: 3.1mm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod.
15532215 Magnification: 10X; Numerical Aperture: 0.4; Working distance: 3.1mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Produktkod. 15532215 Leverantör EVOS™ Leverantörsnummer AMEP4753
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
Designed for demanding research applications
  • Has long working distance (LWD) objective that is optimized for imaging of slides, cell culture dishes and flasks, and microtiter plates
  • Offers Apo correction from UV through IR ranges
  • Parfocal for both visible and IR imaging
  • Coverslip-corrected objective that is optimized for imaging through #1.5 coverslips
  • Compatible with EVOS Cell Imaging systems
  • Offers optical performance from visible light to near infrared light
  • Meets needs across spectrum of magnifications and optical specifications
TRUSTED_SUSTAINABILITY
Specifikationer
Typ Objective Lens
Produktlinje EVOS
För användning med (utrustning) EVOS™ XL Core Imaging System, EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System, EVOS™ FL Auto Imaging System
Inkluderar Vessels, Slides, Cell Culture Dishes, Flasks, Microtiter Well Plates
Fokusavstånd 3.1 mm
Linstyp Super-apochromat
Förstoring 10X
Numerisk bländare 0.4
Kvantitet Each