Läs mer
Omarigliptin, MedChemExpress
Produktkod. 16486948
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
2mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16486948 10 mg -
16480266 2 mg 2mg
16477026 100 mg -
16463007 50 mg -
16429497 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16486948 Leverantör Medchem Express Leverantörsnummer HY15981
MedChemExpress Omarigliptin(MK-3102) is a potent, selective and long-acting DPP-4 inhibitor with IC50 of 1.6 nM; highly selective over all proteases tested (IC50 > 67 (mu)M).
Omarigliptin(MK-3102) is a potent, selective and long-acting DPP-4 inhibitor with IC50 of 1.6 nM
Specifikationer
Kemiskt namn eller material Omarigliptin
CAS 1226781-44-7
Färg Off-White
Anteckningar om renhetsgrad Research
Molekylformel C17H20F2N4O3S
Kvantitet 10 mg
Synonym MK-3102
Löslighetsinformation DMSO : 50 mg/mL (125.49 mM; Need ultrasonic)
LEDER N[C@@H]1[C@@H](C2=CC(F)=CC=C2F)OC[C@H](N3CC4=NN(S(=O)(C)=O)C=C4C3)C1
Molekylvikt (g/mol) 398.43
Formel vikt 398.43
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Metabolism-sugar/lipid metabolism