Läs mer
Omaveloxolone, MedChemExpress
Produktkod. 16485018
Encompass_Preferred
Change view
Kvantitet:
5 mg
10 mg
50 mg
100 mg
Förpackningsstorlek:
100mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16485018 10 mg -
16471257 50 mg -
16447747 5 mg -
16405776 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Produktkod. 16485018 Leverantör Medchem Express Leverantörsnummer HY12212
MedChemExpress Omaveloxolone (RTA 408) is an antioxidant inflammation modulator (AIM), which activates Nrf2 and suppresses nitric oxide (NO). Omaveloxolone attenuates osteoclastogenesis by inhibiting STING dependent NF-κb signaling.
Omaveloxolone (RTA 408) is an antioxidant inflammation modulator (AIM), which activates Nrf2 and suppresses nitric oxide (NO). Omaveloxolone attenuates osteoclastogenesis by inhibiting STING dependent NF-κb signaling.
Specifikationer
Kemiskt namn eller material Omaveloxolone
Fysisk form Powder
CAS 1474034-05-3
Färg White
Kvantitet 10 mg
Anteckningar om renhetsgrad Research
Molekylformel C33H44F2N2O3
För användning med (applikation) Cancer-programmed cell death
Synonym RTA 408
Löslighetsinformation DMSO : ≥ 100 mg/mL (180.27 mM)
LEDER O=C1C(C#N)=C[C@@]2(C)[C@](CC[C@]([C@@]3(C)[C@@]4([H])[C@@]5([H])[C@@](CCC(C)(C)C5)(NC(C(F)(F)C)=O)CC3)(C)C2=CC4=O)([H])C1(C)C
Molekylvikt (g/mol) 554.71
Formel vikt 554.71
Procent renhet 98.0%