Läs mer
omega-Hydroxy-DEET, TRC
Produktkod. 30135469
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135469 500 mg
30003645 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135469 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH934200500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 72236-22-7
Molekylformel C12H17NO2
Molekylvikt (g/mol) 207.27
Synonym N,N-Diethyl-m-hydroxymethylbenzamide, N,N-Diethyl-3-(hydroxymethyl)benzamide
LEDER OCC1=CC(C(N(CC)CC)=O)=CC=C1
Specifikationer
Kemiskt namn eller material omega-Hydroxy-DEET
Anteckningar om renhetsgrad HPLC
Kvantitet 500 mg
InChI formel InChI=1S/C12H17NO2/c1-3-13(4-2)12(15)11-7-5-6-10(8-11)9-14/h5-8,14H,3-4,9H2,1-2H3
Formel vikt 207.1259
Procent renhet >95%