missing translation for 'onlineSavingsMsg'
Läs mer

Omega-Muricholic Acid, TRC

Produktkod. 30467781
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30467781 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 30467781 missing translation for 'mfr' Toronto Research Chemicals missing translation for 'supplierNo' TRCM7327605MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material omega-Muricholic Acid
CAS 6830-03-1
Molekylformel C24 H40 O5
Kvantitet 5 mg
Synonym Cholan-24-oic acid','3,6,7-trihydroxy-','(3α,5β,6α,7β)-, 5β-Cholan-24-oic acid','3α,6α,7β-trihydroxy- (8CI), ω-Muricholic acid (6CI,7CI), (3α,5β,6α,7β)-3,6,7-Trihydroxycholan-24-oic acid, 3α,6α,7β-Trihydroxy-5β-cholanic acid
InChI formel InChI=1S/C24H40O5/c1-13(4-7-19(26)27)15-5-6-16-20-17(9-11-23(15,16)2)24(3)10-8-14(25)12-18(24)21(28)22(20)29/h13-18,20-22,25,28-29H,4-12H2,1-3H3,(H,26,27)/t13-,14-,15-,16+,17+,18+,20+,21-,22-,23-,24-/m1/s1
LEDER C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)[C@H](O)[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC-namn (4R)-4-[(3R,5R,6R,7R,8S,9S,10R,13R,14S,17R)-3,6,7-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
Molekylvikt (g/mol) 408.57
Formel vikt 408.2876
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.