Learn More
Nitritex™ Omega Nerva™ Extra Length Nitrile Gloves, Blue
Catalog Number p-7139092
Click to view available options
Size:
Small
Large
X-Large
2X-Large
Extra length for extra protection. Nitritex™ Omega Nerva™ Extra Length Nitrile Gloves, Blue is a non-sterile, ambidextrous glove packed in an easy access dispenser box that is ideal for use within CSS departments and when handling hazardous chemicals in a laboratory environment.
  • Extra length (400mm/16 in.) provides coverage for the entire forearm
  • Resistant to a wide range of chemicals
  • Textured surface for enhanced grip
  • Powder-free and latex-free
  • Easy-access 100-glove dispenser box packaging
Certifications/Compliance Glove Standard EN 420:2003 & EN 374 (Parts 1, 2 & 3:2003)
Classification Class I Medical & Cat 3 PPE
Color Blue
Disposable Single-use
Exterior Finish Fully Textured
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Examination Gloves
Size X-Large
Sterility Non-sterile
For Use With (Application) CSS departments, hazardous chemicals lab
Glove Cuff Style Beaded/Rolled
Model Omega Nerva
Packaging Type Dispenser box
Puncture Rating 1.5 AQL
Quantity 100/Bx
Resistant To A broad range of chemicals
Series ONAB
Length (English) 16 in.
Length (Metric) 400 mm