missing translation for 'onlineSavingsMsg'
Learn More

Nitritex™ Omega Nerva™ Extra Length Nitrile Gloves, White

Catalog Number 15568721
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Extra length for extra protection. Nitritex™ Omega Nerva™ Extra Length Nitrile Gloves, White is a non-sterile, ambidextrous glove packed in an easy access dispenser box that is ideal for use within CSS departments and when handling hazardous chemicals in a laboratory environment.

  • Extra length (400mm/16 in.) provides coverage for the entire forearm
  • Resistant to a wide range of chemicals
  • Textured surface for enhanced grip
  • Powder-free and latex-free
  • Easy-access 100-glove dispenser box packaging
Certifications/Compliance Glove Standard EN 420:2003 & EN 374 (Parts 1, 2 & 3:2003)
Classification Class I Medical & Cat 3 PPE
Color White
Disposable Single-use
Exterior Finish Fully Textured
Hand Ambidextrous
Latex Free Yes
Material Nitrile
Powdered Powder Free
Product Type Examination Gloves
Size 2X-Large
Sterility Non-sterile
For Use With (Application) CSS departments, hazardous chemicals lab
Glove Cuff Style Beaded/Rolled
Model Omega Nerva
Packaging Type Dispenser box
Puncture Rating 1.5 AQL
Quantity 10 Bx of 100 Ea
Resistant To A broad range of chemicals
Series ONAB
Length (English) 16 in.
Length (Metric) 400 mm
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'