Läs mer
Omega-Tridecenoic Acid, TRC
Produktkod. 30240876
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30240876 5 mg
30240578 10 mg
30240809 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30240876 Leverantör Toronto Research Chemicals Leverantörsnummer TRCT2948455MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6006-06-0
Molekylformel C13H24O2
Molekylvikt (g/mol) 212.33
Synonym (10-Undecenyl)acetic Acid
LEDER OC(=O)CCCCCCCCCCC=C
Specifikationer
Kemiskt namn eller material omega-Tridecenoic Acid