Läs mer
Omeprazole sulfide N-Oxide, TRC
Produktkod. 30009397
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009397 100 mg
30007054 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30009397 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM263040100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 142885-92-5
Molekylformel C17H19N3O3S
Molekylvikt (g/mol) 345.42
Synonym 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-2-pyridinyl)methyl]thio]-1H-benzimidazole, 5-Methoxy-2-[[(4-methoxy-3,5-dimethyl-1-oxo-2-pyridinyl)methyl]sulfanyl]-1H-benzimidazole,
IUPAC-namn 5-methoxy-2-[(4-methoxy-3,5-dimethyl-1-oxidopyridin-1-ium-2-yl)methylsulfanyl]-1H-benzimidazole
LEDER COc1ccc2[nH]c(SCc3c(C)c(OC)c(C)c[n+]3[O-])nc2c1
Specifikationer
Kemiskt namn eller material Omeprazole sulfide N-Oxide
Kvantitet 100 mg
InChI formel InChI=1S/C17H19N3O3S/c1-10-8-20(21)15(11(2)16(10)23-4)9-24-17-18-13-6-5-12(22-3)7-14(13)19-17/h5-8H,9H2,1-4H3,(H,18,19)
Formel vikt 345.1147