Läs mer
Omeprazole Sulfone N-Oxide, TRC
Produktkod. 30409055
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30409055 5 mg
1 options
Produktkod. 30409055 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO6350305MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Omeprazole Sulfone N-Oxide
CAS 158812-85-2
Molekylformel C17 H19 N3 O5 S
Kvantitet 5 mg
Synonym 4-Methoxy-2-[[(5-methoxy-1H-benzimidazol-2-yl)sulphonyl]methyl]-3,5-dimethylpyridine 1-Oxide, Omeprazole Imp. I (EP), Omeprazole Sulphone N-Oxide
InChI formel InChI=1S/C17H19N3O5S/c1-10-8-20(21)15(11(2)16(10)25-4)9-26(22,23)17-18-13-6-5-12(24-3)7-14(13)19-17/h5-8H,9H2,1-4H3,(H,18,19)
LEDER COc1ccc2[nH]c(nc2c1)S(=O)(=O)Cc3c(C)c(OC)c(C)c[n+]3[O-]
IUPAC-namn 5-methoxy-2-[(4-methoxy-3,5-dimethyl-1-oxidopyridin-1-ium-2-yl)methylsulfonyl]-1H-benzimidazole
Molekylvikt (g/mol) 377.41
Formel vikt 377.1045