Learn More
Omi/HtrA2 Protease Inhibitor, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar J65767.MA
Additional Details : Weight : 0.00010kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- A cell-permeable, furfurylidine-thiobarbituric acid compound that acts as a potent, specific, competitive, and reversible inhibitor of the pro-apoptotic, heat-inducible, mitochondrial serine protease, Omi/HtrA2
Specifications
Orange | |
C27H17N3O5S | |
High Temperature Requirement A2 Inhibitor I; 5-((5-(2-Nitrophenyl)furan-2-yl)methylene)-1,3-diphenyl-2-thioxodihydropyrimidine-4,6(1H,5H)-dione | |
Soluble in DMSO at 5mg/ml | |
Solid | |
10 mg |
Protease Inhibitor | |
495.50g/mol | |
Hygroscopic | |
2°C to 8°C | |
A cell-permeable, furfurylidine-thiobarbituric acid compound that acts as a potent, specific, competitive, and reversible inhibitor of the pro-apoptotic, heat-inducible, mitochondrial serine protease, Omi/HtrA2 |