Läs mer
- Home
- Products
- Kromatografikolonner och patroner
- LC-kolonner med lågt till medelhögt tryck
- Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Adjustable Endpieces
Diba Omnifit™ EZ SolventPlus™ Chromatography Columns with Two Adjustable Endpieces
Produktkod. 15895494
Change view
Längd (metrisk):
50 mm
100 mm
150 mm
250 mm
330 mm
400 mm
500 mm
Diameter (metrisk) Inre:
6.6 mm
10 mm
15 mm
25 mm
35 mm
50 mm
Kolumnkapacitet:
0.1 to 0.7 mL
0.1 to 2.4 mL
0.1 to 4.1 mL
0.2 to 5.5 mL
0.2 to 9.4 mL
0.4 to 12.4 mL
0.4 to 21.2 mL
1.2 to 34.4 mL
1.2 to 58.9 mL
10.6 to 38.9 mL
103.1 to 181.6 mL
11 to 23.6 mL
120.2 to 440.7 mL
134.7 to 288.6 mL
152.2 to 230.7 mL
16.5 to 29.1 mL
2.1 to 7.5 mL
2.3 to 115.5 mL
202.0 to 356.0 mL
24.3 to 36.9 mL
24.7 to 53.0 mL
27.7 to 211.7 mL
280.4 to 600.9 mL
29.5 to 108.0 mL
37.1 to 65.4 mL
4.7 to 17.3 mL
4.8 to 10.3 mL
420.6 to 741.1 mL
54.8 to 83.1 mL
620.9 to 941.4 mL
68.7 to 147.3 mL
7.2 to 12.7 mL
Förpackningsstorlek:
Varje
32 product options available for selection
| Produktkod. | Längd (metrisk) | Diameter (metrisk) Inre | Kolumnkapacitet | Förpackningsstorlek |
|---|---|---|---|---|
| 15895494 | 400 mm | 35 mm | 202.0 to 356.0 mL | Varje |
| 15805434 | 330 mm | 6.6 mm | 4.8 to 10.3 mL | Varje |
| 15805464 | 250 mm | 15 mm | 10.6 to 38.9 mL | Varje |
| 15805494 | 150 mm | 35 mm | 2.3 to 115.5 mL | Varje |
| 15885474 | 250 mm | 25 mm | 29.5 to 108.0 mL | Varje |
| 15855444 | 330 mm | 10 mm | 11 to 23.6 mL | Varje |
| 15855474 | 150 mm | 25 mm | 1.2 to 58.9 mL | Varje |
| 15855504 | 330 mm | 50 mm | 280.4 to 600.9 mL | Varje |
| 15885504 | 400 mm | 50 mm | 420.6 to 741.1 mL | Varje |
| 15895434 | 150 mm | 10 mm | 0.2 to 9.4 mL | Varje |
| 15895464 | 500 mm | 15 mm | 54.8 to 83.1 mL | Varje |
| 15815424 | 100 mm | 6.6 mm | 0.1 to 2.4 mL | Varje |
| 15815454 | 500 mm | 10 mm | 24.3 to 36.9 mL | Varje |
| 15815484 | 330 mm | 25 mm | 68.7 to 147.3 mL | Varje |
| 15815514 | 500 mm | 50 mm | 620.9 to 941.4 mL | Varje |
| 15825444 | 250 mm | 10 mm | 4.7 to 17.3 mL | Varje |
| 15825474 | 100 mm | 25 mm | 1.2 to 34.4 mL | Varje |
| 15825504 | 250 mm | 50 mm | 120.2 to 440.7 mL | Varje |
| 15835434 | 400 mm | 6.6 mm | 7.2 to 12.7 mL | Varje |
| 15835464 | 330 mm | 15 mm | 24.7 to 53.0 mL | Varje |
| 15835494 | 250 mm | 35 mm | 27.7 to 211.7 mL | Varje |
| 15845424 | 150 mm | 6.6 mm | 0.1 to 4.1 mL | Varje |
| 15845454 | 100 mm | 15 mm | 0.4 to 12.4 mL | Varje |
| 15845484 | 400 mm | 25 mm | 103.1 to 181.6 mL | Varje |
| 15865434 | 100 mm | 10 mm | 0.2 to 5.5 mL | Varje |
| 15865464 | 400 mm | 15 mm | 37.1 to 65.4 mL | Varje |
| 15865494 | 330 mm | 35 mm | 134.7 to 288.6 mL | Varje |
| 15875424 | 250 mm | 6.6 mm | 2.1 to 7.5 mL | Varje |
| 15875454 | 150 mm | 15 mm | 0.4 to 21.2 mL | Varje |
| 15875484 | 500 mm | 25 mm | 152.2 to 230.7 mL | Varje |
| 15885414 | 50 mm | 6.6 mm | 0.1 to 0.7 mL | Varje |
| 15885444 | 400 mm | 10 mm | 16.5 to 29.1 mL | Varje |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 32 options available.
Denna artikel kan inte returneras.
Se returpolicy
Produktkod.
15895494
Leverantör
Diba
Leverantörsnummer
006EZS3540AA
Logga in eller Skapa webbkonto för att se ditt pris och lagersaldo.
Beräknad leverans: 09-02-2027
Logga inför att se lagersaldo.
Logga inför att se lagersaldo.
Lägg i varukorgen
Lägg i varukorgen
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
- PEEK™ column threads offer outstanding durability
- Ergonomically designed adjustable endpiece offers quick and easy height adjustment
- Superior frit placement adds bed support and even distribution of sample flow
- Reusable columns let you pack your own media specific to your application
Specifikationer
Specifikationer
| Max. Tryck | 150 psi |
| Porstorlek | 10 and 30 μm |
| Kolumnkapacitet | 202.0 to 356.0 mL |
| Längd (metrisk) | 400 mm |
| Material | Borosilicate Glass, PTFE, Chemraz™ |
| Max. Temperatur | 40°C |
| Inkluderar | Two (0.5 m and 2 m L) 1/16 in. OD PTFE tubing assembly with 1/4-28 UNF ETFE fitting at one end, a frit kit (contains two each of 10 μm PE, 10 μm PTFE, and 30 μm PTFE frits) and a Click-N-Seal™ 10-32 PEEK conical port fitting. |
| Temperaturområde (metriskt). | 4 to 40°C |
| Typ | Liquid Chromatography Column |
| Diameter (metrisk) Inre | 35 mm |