Learn More
Diba Omnifit™ Packing Sleeves
Sleeves to connect glass columns
POA
Products 6
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
Product Code Brand Diameter (Metric) Inner Price Quantity & Availability  
15254509
View Documents Promotion Details
Diba
006PS-03
3.0 mm (Bore Size) Columns
POA
Each
View alternative products
Save Up to 
»
N/A
This item requires a price update prior to being ordered.
Request a Price for this item here.
15274509
View Documents Promotion Details
Diba
006PS-10
10 mm (Bore Size) Columns This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15264509
View Documents Promotion Details
Diba
006PS-06
6.6 mm (Bore Size) Columns This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15284509
View Documents Promotion Details
Diba
006PS-15
15 mm (Bore Size) Columns This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15294509
View Documents Promotion Details
Diba
006PS-25
25 mm (Bore Size) Columns This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15204519
View Documents Promotion Details
Diba
006PS-50
50 mm (Bore Size) Columns This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • For Omnifit glass low/medium pressure liquid chromatography columns
  • Offered in multiple I.D. (bore) sizes