Learn More
Kinesis™ Omnifit™ Replacement Filters for Glass Bottles
Solvent frits and filters for the bottom of the bottle remove even small particulates
Brand:  Kinesis™ 009FE-32
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 15224589
  • 620.00 SEK / Pack of 20
Estimated Shipment
04-11-2020
Add to basket
Description
Description
  • All-PTFE construction enables the filters to be used with harsh chemicals
  • The filter units have slots in the body to enable virtually all liquid to be removed from the bottle, minimizing waste
Specifications
Specifications
Replacement Filter
20/Pk.
009F-32
10 μm
PTFE