Learn More
Kinesis™ Omnifit™ Replacement Filters for Glass Bottles
Solvent frits and filters for the bottom of the bottle remove even small particulates
Brand:  Kinesis™ 009FE-45
Additional Details : Weight : 0.40000kg
 View more versions of this product
Product Code. 15244589
  • 811.63 SEK / Pack of 20
Estimated Shipment date
from Supplier 02-08-2019
Add to basket
Description
Description
  • All-PTFE construction enables the filters to be used with harsh chemicals
  • The filter units have slots in the body to enable virtually all liquid to be removed from the bottle, minimizing waste
Specifications
Specifications
0.31cm
20/Pk.
Replacement Filter
GL45 bottle
10μm
PTFE
To be used with the harsh chemicals