missing translation for 'onlineSavingsMsg'
Learn More

Kinesis™ Omnifit™ Replacement Glass Chromatography Columns

Compatible with BenchMark™, SolventPlus™ or HiT™ components

Brand:  Kinesis™ 006RG-15-25

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 15234529

  • 1390.00 SEK / Each


Estimated Shipment
13-11-2020
Add to basket

Description

Description

  • Modular approach allows long life and flexibility in use for Omnifit chromatography components
  • Does not include frits or endpieces

Specifications

Specifications

15 mm
Replacement Glass Chromatography Column
250 mm
Videos
SDS
Documents