missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Onatasertib, MedChemExpress
Beskrivning
Onatasertib (CC-223) is a potent, selective, and orally bioavailable inhibitor of mTOR kinase, with an IC50 value for mTOR kinase of 16 nM. Onatasertib inhibits both mTORC1 and mTORC2.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Onatasertib |
| Fysisk form | Solid |
| CAS | 1228013-30-6 |
| Färg | Light Yellow |
| Kvantitet | 10 mg |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C21H27N5O3 |
| För användning med (applikation) | Cancer-Kinase/protease |
| Synonym | CC-223 ATG-008 |
| Löslighetsinformation | DMSO : ≥ 27 mg/mL (67.93 mM) |
| Visa mer |
Research purposes only
Produkttitel
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?