Learn More
Oncomine™ Gene Browser, Academic License
Oncomine™ Gene Browser, Academic License
Brand:  Invitrogen A18582
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 15514384
  • 58900.00 SEK / Each
Estimated Shipment date
from Supplier 04-04-2018
Add to basket
Oncomine™ Gene Browser, Academic, is a web-based software tool for academic researchers that provides easy access to a comprehensive gene profile across thousands of cancer patient genomes. The rich dataset is presented in a visual and dynamic environment that easily conveys significant gene mutations, expression, and DNA copy number changes. It also enables customers to quickly and easily ask questions across hundreds of cancer subtypes for any gene. Designed for a broad audience, the Oncomine™ Gene Browser is fast, easy to use, and provides powerful insight into any gene target by deriving knowledge from the world’s oncology experiments.
  • Profile your critical targets and biomarkers across tens of thousands of retrospective de-identified patient tumor genomes online, in minutes
  • Rapidly prioritize targets based on mutations, copy number variants, and differential expression
  • Identify optimal patient segments for your clinical development based on biomarker frequencies in well-characterized patient populations
    A key benefit of Oncomine™ Gene Browser is the fast and comprehensive evaluation of your oncology gene targets within the world's oncology experimental data. No longer relying on one or two research publications for validation or being restricted to author interpretation and focus, an unbiased approach to evaluating your gene targets can be performed in minutes, often yielding insights into new cancer subtypes that can expand your target clinical population.
    Oncomine™ Gene Browser empowers users to assess mutations, gene expression, DNA copy number, cancer subtypes, normal tissue expression, and pick cell line models. In a nutshell, millions of dollars worth of experiments are at your fingertips and can be 'performed' in silico so that you can design and execute fewer, more focused wet lab experiments on the targets and indications that have a greater chance for success.
    Learn more about Oncomine™ Gene Browser.
    Oncomine™ Gene Browser, Academic, can be licensed for various user numbers, up to an institutional license with an unlimited number of registered users.
    Note: This product is for academic and nonprofit cancer researchers only. If you are a commercial/for-profit cancer researcher, please contact one of our knowledgable sales specialists at compendiasales@lifetech.com.
    How to Order
    Order this product as you would any standard product. You will receive an email with your user name and temporary password, which you can then use to log in and get started.
  • Specifications
    Oncomine™ Academic License Gene Browser
    1 Each
    Oncomine™ Academic License Gene Browser
    Cancer genomics
    Oncomine™ Academic License Gene Browser, 1 Year, 1 User, Academic, Annual, Single User License Type, The Rich Dataset is Presented in a Visual and Dynamic Environment that Easily Conveys Significant Gene Mutations, Expression and DNA Copy Number Cha
    1 Year, 1 User
    For research use only
    For Research Use Only. Not for use in diagnostic procedures.