Läs mer
Tocris Bioscience™ Ondansetron hydrochloride
GREENER_CHOICE
Produktkod. 18782554
Change view
Kvantitet:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18782554 10 mg
18722704 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 18782554 Leverantör Tocris Bioscience™ Leverantörsnummer 2891/10
Selective 5-HT3 antagonist
Ondansetron hydrochloride is a selective 5-HT3 receptor antagonist (Ki = 6.16 nM). Also an inhibitor of human multidrug and toxin extrusion (MATE) transporter 1 (IC50 = <10-160 nM). Ondansetron blocks 5-HT-evoked transient inward currents (IC50 = 0.1 nM) in human recombinant h5-HT3A receptors. In an animal model of Parkinson's disease, Ondansetron reduces both the severity of dyskinesia and psychosis-like behaviors. Antiemetic; prevents emesis induced by cytotoxic drugs and radiation.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 99614-01-4
Molekylformel C18H20ClN3O
Specifikationer
Kemiskt namn eller material Ondansetron hydrochloride
Kvantitet 10 mg
Formel vikt 329.83
Renhet >99%
Molekylvikt (g/mol) 329.83