Läs mer
Tocris Bioscience™ ONO 8130
EP1 receptor antagonist
Brand: Tocris Bioscience™ 5406/50
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
ONO 8130 is an EP1 receptor antagonist. Exhibits analgesic effect in a mouse mode of cystitis. Orally bioavailable.Specifikationer
| ONO 8130 | |
| 4-[[[2,3-Dihydro-6-[(2-methylpropyl)[(4-methyl-2-thiazolyl)sulfonyl]amino]-1H-indene-5yl]oxy]methyl]benzoic acid | |
| Soluble to 100mM in DMSO and to 20mM in ethanol | |
| Bioactive Small Molecule |
| 459841-96-4 | |
| 50 mg | |
| Solid |
Produkttitel
Research Use Only. Not For Human or Veterinary Use