missing translation for 'onlineSavingsMsg'
Läs mer

ONO-8590580, MedChemExpress

Produktkod. 16467417
Encompass_Preferred
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
10 mg
25 mg
50 mg
100 mg
Förpackningsstorlek:
25mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
16467417 5 mg -
16453206 25 mg 25mg
16430997 50 mg -
16424708 10 mg -
16405536 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 16467417 Leverantör Medchem Express Leverantörsnummer HY112788

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

MedChemExpress ONO-8590580 is a GABAA α5 negative allosteric modulator.

ONO-8590580 is a GABAA α5 negative allosteric modulator.

Specifikationer

Kemiskt namn eller material ONO-8590580
CAS 1802661-73-9
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C21H21FN6
Kvantitet 5 mg
Löslighetsinformation DMSO : 62.5 mg/mL (166.03 mM; Need ultrasonic)
LEDER CN1C=C(C2=CN=C(NC3=C(F)C(C)=C4N=CN(CC5CC5)C4=C3)C=C2)N=C1
Molekylvikt (g/mol) 376.43
Formel vikt 376.43
Procent renhet 99.13%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Neuroscience-Neuromodulation
Visa mer Visa mindre

Research purposes only

Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.