Beskrivning
- Protects environment from droplets emitted by wearer
- Effective against bacteria, particles, and viruses
- Feeldry and Softex inner layer absorbs moisture to prevent skin reactions
- New easy take out method (ties are folded under each mask for convenience)
- Intended for healthcare professionals and in operating room to protect against splashes
Specifikationer
Specifikationer
| Färg | Blått |
| Fästtyp | Slipsar |
| Material | Polypropen |
| Certifieringar/efterlevnad (Europa) | (EU) 2017/745, EN 14683, ISO 9001, ISO 13485 |
| Disponibel | Engångsbruk |
| Latexfri | Ja |
| Förpackningsmängd | 50 Pk. |