Learn More
Grant Instruments™ Optima Bridge Plate
View and access thermostat on heating and cooling circulators. Grant Instruments™ Optima Bridge Plate is for use with Grant refrigeration units, heating and cooling circulators.
842.70 SEK - 1945.10 SEK
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12757799
|
Grant Instruments™
T-BPS |
T Optima Heating Circulator models to P5 |
1749.00 SEK
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
12707799
|
Grant Instruments™
T-BTL |
T Optima Heating Circulator models to ST12, 18, 26 and 38 |
1945.10 SEK
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
12717799
|
Grant Instruments™
T-BSS |
T Optima Heating Circulator S5 tank |
1722.50 SEK
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
12797789
|
Grant Instruments™
T-BTS |
T Optima Heating Circulator ST5 tank |
1722.50 SEK
Each |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
12747789
|
Grant Instruments™
T-BR5 |
T Optima Heating Circulator models to R5 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12727799
|
Grant Instruments™
T-BSL |
T Optima heating circulator models to S12, 18, 26, 38 tanks |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12767799
|
Grant Instruments™
T-BPL |
T Optima Heating Circulator models to P12 and P18 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12727789
|
Grant Instruments™
T-BR3 |
T Optima Heating Circulator models to R1, R2, R3 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
12737789
|
Grant Instruments™
T-BR4 |
T Optima Heating Circulator models to R4 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
View and access thermostat on heating and cooling circulators. Grant Instruments™ Optima Bridge Plate is for use with Grant refrigeration units, heating and cooling circulators.
- For use with Grant refrigeration units, heating and cooling circulators
- Ensures visibility/accessibility of the thermostat whilst optimising bench space