Läs mer
SHP Steriltechnik AG Option V Laboklav 55-195, Vacuum
Beskrivning
- Permanent or fractional pre-vacuum for optimal air removal
- Reliable penetration of steam into sterilisation items that are difficult to vent
- Fractional pre-vacuum ensures reproducible sterilisation, DIN 58951-2
- Drying vacuum for efficient post-drying of packaged items
- Condensate is reliably removed during the drying phase
- Avoids the need for additional transfer to a drying cabinet
- Suitable for solid, moist or packaged items
Specifikationer
Specifikationer
| För användning med (utrustning) | Autoclaves, Laboklav 55-195 |
| Modell | Option V Laboklav 55-195 |
| Produkttyp | Autoclave Vacuum |