Läs mer
OR-486, TRC
Produktkod. 30419880
Change view
Kvantitet:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30419880 500 mg
30418112 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30419880 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO670500500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 7659-29-2
Molekylformel C6 H4 N2 O6
Molekylvikt (g/mol) 200.11
Synonym 3,5-Dinitrobenzene-1,2-diol, Entacapone Imp. E (EP)
IUPAC-namn 3,5-dinitrobenzene-1,2-diol
LEDER Oc1cc(cc(c1O)[N+](=O)[O-])[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material 3,5-Dinitrobenzene-1,2-diol
Kvantitet 500 mg
InChI formel InChI=1S/C6H4N2O6/c9-5-2-3(7(11)12)1-4(6(5)10)8(13)14/h1-2,9-10H
Formel vikt 200.0069