missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OR1S1/OR1S2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA561927
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FSSFLQIGRN MHQGNQTTIT EFILL Highest antigen sequence identity to the following orthologs: Mouse - 56%, Rat - 52%.
Olfactory receptors interact with odorant molecules in the nose, to initiate a neuronal response that triggers the perception of a smell. The olfactory receptor proteins are members of a large family of G-protein-coupled receptors (GPCR) arising from single coding-exon genes. Olfactory receptors share a 7-transmembrane domain structure with many neurotransmitter and hormone receptors and are responsible for the recognition and G protein-mediated transduction of odorant signals. The olfactory receptor gene family is the largest in the genome. The nomenclature assigned to the olfactory receptor genes and proteins for this organism is independent of other organisms.Specifications
OR1S1/OR1S2 | |
Polyclonal | |
Unconjugated | |
OR1S1 | |
Olfactory receptor 1S1; olfactory receptor 1S2; olfactory receptor family 1 subfamily S member 1 (gene/pseudogene); olfactory receptor family 1 subfamily S member 2; olfactory receptor OR11-231; Olfactory receptor OR11-232; olfactory receptor, family 1, subfamily S, member 1; olfactory receptor, family 1, subfamily S, member 2; OR11-231; OR11-232; OR1S1; OR1S2; OST034 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
219958, 219959 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8NGQ3, Q8NH92 | |
OR1S1, OR1S2 | |
Recombinant protein corresponding to Human OR1S1/OR1S2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only