missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ ORC4L (Human) Recombinant Protein

Human ORC4L full-length ORF ( AAH05388, 1 a.a. - 248 a.a.) recombinant protein with GST-tag at N-terminal.

Brand:  Abnova™ H00005000-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16138611

  • 4515.00 SEK / 10µg
Estimated Shipment: 29-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Sequence: MSSRKSKSNSLIHTECLSQVQRILRERFCRQSPHSNLFGVQVQYKHLSELLKRTALHGESNSVLIIGPRGSGKTMLISHALKELMEIEEVSENVLQVHLNGLLQINDKIALKEITRQLNLENVVGDKVFGSFAENLSFLLEALKKGDRTSSCPVIFILDEFDLFAHHKNQTLLYNLFDISQSAQTPIAVIGLTCRLDILELLEKRVKSRFSHRQIHLMNSFGFPQYVKIFKEQLSLPAEFPDKVFAEK
Specifications

Specifications

AAH05388
origin recognition complex, subunit 4-like (yeast)
125% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ORC4L
GST
5000
Wheat germ expression system
10 μg
ORC4, ORC4P
Wheat Germ (in vitro)
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers