missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORC5L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | ORC5L |
---|---|
Concentration | 0.23 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16838693
|
Proteintech
11542-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16828693
|
Proteintech
11542-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 03-04-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Component of the origin recognition complex (ORC) that binds origins of replication. DNA-binding is ATP-dependent. The specific DNA sequences that define origins of replication have not been identified yet. ORC is required to assemble the pre- replication complex necessary to initiate DNA replication.Specifications
ORC5L | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O43913, Q9WUV0 | |
26429, 362304, 5001 | |
ORC5L Fusion Protein Ag2133 | |
Primary | |
-20°C | |
ORC5 |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
ORC5, ORC5L, ORC5P, ORC5T | |
Orc5 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |