missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ORC5L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11542-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Component of the origin recognition complex (ORC) that binds origins of replication. DNA-binding is ATP-dependent. The specific DNA sequences that define origins of replication have not been identified yet. ORC is required to assemble the pre- replication complex necessary to initiate DNA replication.Specifications
ORC5L | |
Polyclonal | |
Unconjugated | |
ORC5 | |
ORC5, ORC5L, ORC5P, ORC5T | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
26429, 362304, 5001 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O43913, Q9WUV0 | |
Orc5 | |
ORC5L Fusion Protein Ag2133 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |