missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Orcinol, 99.6%, MP Biomedicals™
Click to view available options
Quantity:
5 g
10 g
50 g
Spezifikation
Color | Tan to White |
Assay Percent Range | ∽ 98% |
Quantity | 5 g |
Formula Weight | 142.2 |
Percent Purity | 99.6% |
Physical Form | Powder |
Chemical Name or Material | Orcinol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'