missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oregon Pesticide Mixture 476 600 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Kemiskt namn eller material | (E)-Fenpyroximate∣Acequinocyl∣Acetamiprid∣Azoxystrobin∣Bifenazate∣Boscalid∣Chlorfenapyr∣Etoxazole∣Fludioxonil∣Imidacloprid∣Kresoxim-methyl∣Metalaxyl∣MGK 264∣Piperonyl butoxide∣Spiromesifen∣Spirotetramat∣Spiroxamine∣Trifloxystrobin |
| CAS | 134098-61-6, 57960-19-7, 135410-20-7, 131860-33-8, 149877-41-8, 188425-85-6 |
| Analyt- eller komponentnamn | (E)-Fenpyroximate∣Acequinocyl∣Acetamiprid∣Azoxystrobin∣Bifenazate∣Boscalid∣Chlorfenapyr∣Etoxazole∣Fludioxonil∣Imidacloprid∣Kresoxim-methyl∣Metalaxyl∣MGK 264∣Piperonyl butoxide∣Spiromesifen∣Spirotetramat∣Spiroxamine∣Trifloxystrobin |
| Frakt skick | Room Temperature |
| Certifikat och efterlevnad | ISO 17034 |
| Lösningsmedel eller matris | Acetonitrile |
| Kvantitet | 1 ml |
| Koncentration eller sammansättning (efter analyt eller komponenter) | (E)-Fenpyroximate 600 μg/mL∣Acequinocyl 600 μg/mL∣Acetamiprid 600 μg/mL∣Azoxystrobin 600 μg/mL∣Bifenazate 600 μg/mL∣Boscalid 600 μg/mL∣Chlorfenapyr 600 μg/mL∣Etoxazole 600 μg/mL∣Fludioxonil 600 μg/mL∣Imidacloprid 600 μg/mL∣Kresoxim-methyl 600 μg/mL∣Metalaxyl 600 μg/mL∣MGK 264 600 μg/mL∣Piperonyl butoxide 600 μg/mL∣Spiromesifen 600 μg/mL∣Spirotetramat 600 μg/mL∣Spiroxamine 600 μg/mL∣Trifloxystrobin 600 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?