Beskrivning
ORF8 Monoclonal antibody specifically detects ORF8 in SARS-CoV-2 samples. It is validated for Immunohistochemistry,Western Blot,Immunocytochemistry
Specifikationer
Specifikationer
| Antigen | ORF8 |
| Användningsområden | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Klassificering | Monoclonal |
| Klona | 1041422 |
| Konjugera | PE |
| Formulering | PBS |
| Värd art | Mouse |
| Immunogen | E. coli-derived human SARS-CoV-2 ORF8, Met16-His121, Accession # YP_009724369 |
| Reningsmetod | Protein A or G purified from hybridoma culture supernatant |
| Kvantitet | 0.1 mL |
| Visa mer |