Läs mer
Thermo Scientific™ Orion™ Single Junction Refillable Epoxy Bodied Combination pH Electrode
Kontakttyper:
BNC vattentät, RCA
Skruvlock
Förpackningsstorlek:
Varje
Beskrivning
Thermo Scientific™ Orion™ Silver Chloride Refillable Electrodes provide reliable and accurate measurements of routine samples. They are available is epoxy bodies for ruggedness and best suited for general purpose samples and drinking water.
Includes:
- 60ml bottle of Orion electrode fill solution, 900011
Specifikationer
Specifikationer
| Beskrivning | With screw cap connector |
| Kontakttyper | Skruvlock |
| Korsning | Glass Fiber |
| Material | Epoxy Body |
| pH-intervall | 0 to 14 |
| Temperatur (metrisk) Drift | 0°C to 90°C |
| Typ | Combination pH Electrode |
| Typ av fyllningslösning | 4M KCl with Ag/AgCl |
| Intern referens | Ag/AgCl |
| Precision | 0.02 |
| Visa mer |