Läs mer
ORM-15341, TRC
Produktkod. 30427926
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30427926 50 mg
30426873 10 mg
30424292 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30427926 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO41001050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1297537-33-7
Molekylformel C19H17ClN6O2
Molekylvikt (g/mol) 396.83
Synonym ORM-15341, (S)-5-Acetyl-N-(1-(3-(3-chloro-4-cyanophenyl)-1H-pyrazol-1-yl)propan-2-yl)-1H-pyrazole-3-carboxamide
IUPAC-namn 5-acetyl-N-[(2S)-1-[3-(3-chloro-4-cyanophenyl)pyrazol-1-yl]propan-2-yl]-1H-pyrazole-3-carboxamide
LEDER C[C@@H](Cn1ccc(n1)c2ccc(C#N)c(Cl)c2)NC(=O)c3cc([nH]n3)C(=O)C
Specifikationer
Kemiskt namn eller material (S)-5-Acetyl-N-(1-(3-(3-chloro-4-cyanophenyl)-1H-pyrazol-1-yl)propan-2-yl)-1H-pyrazole-3-carboxamide
Kvantitet 50 mg
InChI formel InChI=1S/C19H17ClN6O2/c1-11(22-19(28)18-8-17(12(2)27)23-24-18)10-26-6-5-16(25-26)13-3-4-14(9-21)15(20)7-13/h3-8,11H,10H2,1-2H3,(H,22,28)(H,23,24)/t11-/m0/s1
Formel vikt 396.1102