missing translation for 'onlineSavingsMsg'
Läs mer

Orotic Acid-13C,15N2 Monohydrate, TRC

Produktkod. 30008920
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30008920 10 mg
30004535 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30008920 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO69150210MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 1346602-15-0
Molekylformel 13C C4 H4 15N2 O4 . H2 O
Molekylvikt (g/mol) 177.09
Synonym 1,2,3,6-Tetrahydro-2,6-dioxo-4-pyrimidine-2-13C-1,3-15N2-carboxylic acid monohydrate, Orotic acid-13C,15N2 monohydrate, Orotic acid-2-13C,1,3-15N2 monohydrate, 2,4-Dioxo-1H-pyrimidine-2-13C-1,3-15N2-6-carboxylic acid monohydrate, 2,4-Dihydroxypyrimidine-2-13C-1,3-15N2-6-carboxylic acid monohydrate
IUPAC-namn 2,4-dioxo-(2^{13}C,1,3-^{15}N_{2})1H-pyrimidine-6-carboxylic acid;hydrate
LEDER O.OC(=O)C1=CC(=O)[15NH][13C](=O)[15NH]1

Specifikationer

Kemiskt namn eller material Orotic Acid-13C, 15N2 Monohydrate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C5H4N2O4.H2O/c8-3-1-2(4(9)10)6-5(11)7-3;/h1H,(H,9,10)(H2,6,7,8,11);1H2/i5+1,6+1,7+1;
Formel vikt 177.0251
Procent renhet >95%
Produkttitel
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.