missing translation for 'onlineSavingsMsg'
Learn More

Orotic acid, 98%, anhydrous, Thermo Scientific Chemicals

Catalog Number p-4228748
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
65-86-1
C5H4N2O4
156.1
PXQPEWDEAKTCGB-UHFFFAOYSA-N
orotic acid, 6-carboxyuracil, orodin, oropur, orotonin, orotonsan, oroturic, orotyl, vitamin b13, whey factor
967
CHEBI:16742
2,4-dioxo-1H-pyrimidine-6-carboxylic acid
C1=C(NC(=O)NC1=O)C(=O)O
This item is not returnable. View return policy
65-86-1
C5H4N2O4
156.1
PXQPEWDEAKTCGB-UHFFFAOYSA-N
orotic acid, 6-carboxyuracil, orodin, oropur, orotonin, orotonsan, oroturic, orotyl, vitamin b13, whey factor
967
CHEBI:16742
2,4-dioxo-1H-pyrimidine-6-carboxylic acid
C1=C(NC(=O)NC1=O)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula C4N2H3(O)2COOH
Quantity 10 g
Beilstein 25,253
Merck Index 15,6972
Solubility Information Solubility in water: slightly soluble in water
Formula Weight 156.1
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Orotic acid
Show More Show Less
compliance-icons
Product Identifier
  • Orotic acid, anhydrous, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'