missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Invitrogen™ ORP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA521860
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
Recommended positive controls: A431, H1299, HeLa, HepG2, mouse brain. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the oxysterol-binding protein family, a group of intracellular lipid receptors. Most members contain an N-terminal pleckstrin homology domain and a highly conserved C-terminal OSBP-like sterol-binding domain, although some members contain only the sterol-binding domain. Transcript variants derived from alternative promoter usage and/or alternative splicing exist; they encode different isoforms.
Specifikationer
| ORP1 | |
| Polyclonal | |
| Unconjugated | |
| OSBPL1A | |
| DCDC4A; FLJ10217; FLJ50293; FLJ55454; FLJ79410; G430090F17Rik; Gm753; ORP1; ORP-1; Orp1a; Orp1l; OSBP8; OSBPL1; OSBPL1A; OSBPL1B; OSBP-related protein 1; oxysterol binding protein like 1A; oxysterol binding protein-like 1A; oxysterol binding protein-like 1B; oxysterol-binding protein-related protein 1; oxysterol-binding protein-related protein 1 variant 1; oxysterol-binding protein-related protein 1 variant 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114876, 64291 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
| Q91XL9, Q9BXW6 | |
| OSBPL1A | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ORP1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering