Läs mer
Orphenadrine Hydrochloride, LoGiCal
Produktkod. 30463670
Change view
Kvantitet:
10 mg
Produktkod. Kvantitet
30463670 10 mg
1 options
Produktkod. 30463670 Leverantör LoGiCal Leverantörsnummer LGCFOR0801.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Orphenadrine Hydrochloride
CAS 341-69-5
Analyt- eller komponentnamn Orphenadrine Hydrochloride
Frakt skick Room Temperature
Molekylformel C18 H23 N O . Cl H
Certifikat och efterlevnad ISO 9001
Synonym Orphenadrine Hydrochloride, N,N-Dimethyl-2-[(2-methylphenyl)phenylmethoxy]ethanamine hydrochloride, o-Methyldiphenhydramine hydrochloride, o-Monomethyldiphenhydramine hydrochloride, 2-(Phenyl-o-tolylmethoxy)ethyldimethylamine hydrochloride, Phenyl-o-tolylmethyl dimethylaminoethyl ether hydrochloride, Disipal, Mephenamin, N,N-Dimethyl-2-(o-methyl-alpha-phenylbenzyloxy)ethylamine hydrochloride, beta-Dimethylaminoethyl 2-methylbenzhydryl ether hydrochloride
InChI formel InChI=1S/C18H23NO.ClH/c1-15-9-7-8-12-17(15)18(20-14-13-19(2)3)16-10-5-4-6-11-16;/h4-12,18H,13-14H2,1-3H3;1H
Lösningstyp Neat
LEDER Cl.CN(C)CCOC(c1ccccc1)c2ccccc2C
IUPAC-namn N,N-dimethyl-2-[(2-methylphenyl)-phenylmethoxy]ethanamine;hydrochloride
Industry Type Clinical Testing
Molekylvikt (g/mol) 305.84
Kvantitet 10 mg
Formel vikt 305.1546 g/mol