Läs mer
Oryzalin, TRC
Produktkod. 30425229
Change view
Kvantitet:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425229 250 mg
30421194 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30425229 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO675503250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 19044-88-3
Molekylformel C12 H18 N4 O6 S
Molekylvikt (g/mol) 346.36
Synonym Sulfanilamide, 3,5-dinitro-N4,N4-dipropyl- (8CI), 4-(Dipropylamino)-3,5-dinitrobenzenesulfonamide, 3,5-Dinitro-N4,N4-dipropylsulfanilamide, Dirimal, EL 119, N4,N4-Dipropyl-3,5-dinitrosulfanilamide, NSC 648479, Oryzalin, Oryzalin 4 Pro, Rizelan, Ryzelan, Surflan
IUPAC-namn 4-(dipropylamino)-3,5-dinitrobenzenesulfonamide
LEDER CCCN(CCC)c1c(cc(cc1[N+](=O)[O-])S(=O)(=O)N)[N+](=O)[O-]
Specifikationer
Kemiskt namn eller material Oryzalin
Kvantitet 250 mg
InChI formel InChI=1S/C12H18N4O6S/c1-3-5-14(6-4-2)12-10(15(17)18)7-9(23(13,21)22)8-11(12)16(19)20/h7-8H,3-6H2,1-2H3,(H2,13,21,22)
Formel vikt 346.0947