missing translation for 'onlineSavingsMsg'
Learn More

OSBPL3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12417-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840124

  • 4721.67 SEK / 150µL
Estimated Shipment: 03-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ZNF703 belongs to the Elbow/Noc family. It contains 1 C2H2-type zinc finger.ZNF703 may function as a transcriptional repressor.
Specifications

Specifications

OSBPL3
Polyclonal
Unconjugated
OSBPL3
KIAA0704, ORP 3, ORP3, OSBP related protein 3, OSBP3, OSBPL3
Rabbit
Antigen Affinity Chromatography
RUO
26031, 362360, 71720
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9DBS9, Q9H4L5
Osbpl3
OSBPL3 Fusion Protein Ag3044
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers