missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSCP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1656.73 SEK - 4721.67 SEK
Specifications
Antigen | OSCP1 |
---|---|
Concentration | 0.19 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16850394
|
Proteintech
12598-1-AP-20UL |
20 μL |
1656.73 SEK
20µL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16840394
|
Proteintech
12598-1-AP-150UL |
150 μL |
4721.67 SEK
150µL |
Estimated Shipment: 20-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
The function of this protein remains unknown.Specifications
OSCP1 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q4QQS3, Q8BHW2, Q8WVF1 | |
127700, 230751, 362595 | |
OSCP1 Fusion Protein Ag3209 | |
Primary | |
-20°C | |
OSCP1 |
0.19 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
C1orf102, hOSCP1, NOR1, OSCP1, Protein OSCP1 | |
OSCP1 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |