Läs mer
Oseltamivir Acid Hydrochloride, TRC
Produktkod. 30414126
Change view
Kvantitet:
2.5 mg
10 mg
100 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30414126 100 mg -
30413089 2.5 mg 2.5mg
30411990 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30414126 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO700995100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1415963-60-8
Molekylformel C14 H24 N2 O4 . Cl H
Molekylvikt (g/mol) 320.81
Synonym 1-Cyclohexene-1-carboxylic acid, 4-(acetylamino)-5-amino-3-(1-ethylpropoxy)-, hydrochloride (1:1), (3R,4R,5S)-, Oseltamivir acid hydrochloride, (3R,4R,5S)-4-Acetamido-5-amino-3-(1-ethylpropoxy)cyclohex-1-ene-1-carboxylic acid hydrochloride
IUPAC-namn (3R,4R,5S)-4-acetamido-5-amino-3-pentan-3-yloxycyclohexene-1-carboxylic acid;hydrochloride
LEDER Cl.CCC(CC)O[C@@H]1C=C(C[C@H](N)[C@H]1NC(=O)C)C(=O)O
Specifikationer
Kemiskt namn eller material Oseltamivir acid hydrochloride
Kvantitet 100 mg
InChI formel InChI=1S/C14H24N2O4.ClH/c1-4-10(5-2)20-12-7-9(14(18)19)6-11(15)13(12)16-8(3)17;/h7,10-13H,4-6,15H2,1-3H3,(H,16,17)(H,18,19);1H/t11-,12+,13+;/m0./s1
Formel vikt 320.1503