Läs mer
Oseltamivir Namino-fructosyl Conjugate, TRC
Produktkod. 17937454
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17937454 25 mg
17907714 10 mg
17947734 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17937454 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO70101525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2413185-85-8
Molekylformel C22H38N2O9
Molekylvikt (g/mol) 474.55
IUPAC-namn Ethyl (4R,5S)-4-Acetamido-3-(pentan-3-yloxy)-5-((((3S,4R,5R)-2,3,4,5-tetrahydroxytetrahydro-2H-pyran-2-yl)methyl)amino)cyclohex-1-ene-1-carboxylate
LEDER O=C(OCC)C1=CC(OC(CC)CC)[C@H](NC(C)=O)[C@@H](NCC2(OC[C@@H](O)[C@@H](O)[C@@H]2O)O)C1
Specifikationer
Kemiskt namn eller material Oseltamivir Namino-fructosyl Conjugate
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C22H38N2O9/c1-5-14(6-2)33-17-9-13(21(29)31-7-3)8-15(18(17)24-12(4)25)23-11-22(30)20(28)19(27)16(26)10-32-22/h9,14-20,23,26-28,30H,5-8,10-11H2,1-4H3,(H,24,25)/t15-,16+,17?,18+,19+,20-,22?/m0/s1
Formel vikt 474.26
Procent renhet >95%