Läs mer
Oseltamivir, TRC
Produktkod. 30425331
Change view
Kvantitet:
100 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30425331 1 g
30425040 250 mg
30415052 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30425331 Leverantör Toronto Research Chemicals Leverantörsnummer TRCO7010051G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 196618-13-0
Molekylformel C16 H28 N2 O4
Molekylvikt (g/mol) 312.4
Synonym 1-Cyclohexene-1-carboxylic acid, 4-(acetylamino)-5-amino-3-(1-ethylpropoxy)-, ethyl ester, (3R,4R,5S)-, 1-Cyclohexene-1-carboxylic acid, 4-(acetylamino)-5-amino-3-(1-ethylpropoxy)-, ethyl ester, [3R-(3α,4β,5α)]-, (-)-Oseltamivir, Enfluvir, Ethyl (3R,4R,5S)-5-amino-4-acetamido-3-(pentan-3-yloxy)cyclohex-1-ene-1-carboxylate, GOP-A-Flu, GS 4104, Oseltamivir, Tamiflu-Free, Tamvir, ZINC 039295508, Ethyl (3R,4R,5S)-4-acetamido-5-amino-3-(1-ethylpropoxy)cyclohex-1-ene-1-carboxylate
IUPAC-namn ethyl (3R,4R,5S)-4-acetamido-5-amino-3-pentan-3-yloxycyclohexene-1-carboxylate
LEDER CCOC(=O)C1=C[C@@H](OC(CC)CC)[C@H](NC(=O)C)[C@@H](N)C1
Specifikationer
Kemiskt namn eller material Oseltamivir (free base)
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C16H28N2O4/c1-5-12(6-2)22-14-9-11(16(20)21-7-3)8-13(17)15(14)18-10(4)19/h9,12-15H,5-8,17H2,1-4H3,(H,18,19)/t13-,14+,15+/m0/s1
Formel vikt 312.2049
Procent renhet >95%