missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSM Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27792-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
Oncostatin M is a member of a cytokine family that includes leukemia-inhibitory factor, granulocyte colony-stimulating factor, and interleukin 6. This gene encodes a growth regulator which inhibits the proliferation of a number of tumor cell lines. It regulates cytokine production, including IL-6, G-CSF and GM-CSF from endothelial cells.Specifications
OSM | |
Polyclonal | |
Unconjugated | |
OSM | |
oncostatin M, OSM | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
289747, 5008 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P13725, Q65Z15 | |
Osm | |
OSM Fusion Protein Ag27093 | |
20 μL | |
Primary | |
Rat, Human | |
Antibody | |
IgG |